All-Layer Drill, Burkle
Supplier: BURKLE INC
5310-0150
76501-526EA 2460.3 USD
76501-526
All-Layer Drill, Burkle
Samplers Solid Samplers
The all-layer drill is a zone collector specifically designed for heavily compacted substances.
  • Stainless steel AISI 316 L (1.4404)
  • 25 mm (0.98") diameter
Often it is very difficult for conventional zone collectors to penetrate compacted media and take a sample extending through all layers. By contrast, the drilling blades of the all-layer drill burrow easily and quickly to the required depth without expending any great force. This makes it easy to take samples of media, even those with high density, and every type of highly compacted powders and granulates. The all-layer drill thus allows users to take a representative sample through all the layers, even with greatly compressed media.
Ideal for large containers, mixed containers, big bags, etc. in the building materials, chemical, pharmaceutical, food, and luxury items industries.