Microsart® e.jet Laboratory Liquid Transfer Pump
Supplier: Sartorius
166MP-4
76500-774EA 2114.61 USD
76500-774
Microsart® e.jet Laboratory Liquid Transfer Pump
Pumps Drum/Transfer Pumps
The Microsart® e.jet Fluid Pump is a small, state-of-the-art liquid transfer pump for optimal vacuum filtration in microbiological QC lab.
  • Small foot print - no need of suction flasks between pump and manifold
  • Simultaneous transfer of filtered liquid to waste
  • Constant flow rates and defined maximum vacuum
  • Maximum vacuum: 5.80 psi
  • Pump flow: 4 L/min
The Microsart® e.jet Fluid Pump is a superior choice for your vacuum filtration needs. This laboratory transfer pump is equipped with a quick connection. It is ideal for filtration of liquid samples in microbiological quality control, achieving a transmembrane pressure of 600 mbar and a flow rate of 4.0 NI/min (4.0 Normiliters water displacement by air in one minute).
Smooth and reliable filtration: The Microsart® e.jet fluid pump was designed to be efficient and highly reliable. Constant flow rates and a defined maximum vacuum guarantee smooth and reliable filtration.
Reduced complexity and space: Traditional pumps consist of numerous parts, including connectors, tubes, vacuum containers, protection filters, Woulff’s bottle and the vacuum pump. After several samples, the vacuum must be broken using those traditional pumps to empty the filtrate collection container. These complex systems require more laboratory space and are time-consuming. These needs are compactly addressed with the Microsart® e.jet fluid pump.
The Microsart® e.jet Fluid Pump is ideal for 1, 3, and 6-branch manifolds. Building up the vacuum filtration system is easy and fast, with innovative quick connections.
Certifications: Protection class III. Dust / Water Protection According to IEC 529 IP 64.