Membrane Filters, Type 131, Sartorius
Supplier: Sartorius
1310150AHN
76500-650PK 265.03 USD
76500-650
Membrane Filters, Type 131, Sartorius
Filters Membrane Filters
Non sterile membrane filters for colony counting, particle testing, microscopy and sterility testing. The membrane filters are made of cellulose nitrate (cellulose ester).