Simple Green Clean Building All-Purpose Cleaner Concentrate, Essendant
Supplier: Janitorial Supplies
SMP11001CT
76500-462CT 48.11 USD
76500-462
Simple Green Clean Building All-Purpose Cleaner Concentrate, Essendant
Cleaning Agents Detergents and Surface Cleansers
This biodegradable all-purpose cleaner removes dirt, grime, and oily soils without heavy scrubbing. Strong surfactants penetrate, lift, and remove soils, rinsing cleanly for a streak-free and smudge-free shine.
  • Biodegradable formula
  • Fragrance-free to reduce the impact on indoor air quality
  • Cold water compatible
  • Non-caustic and non-flammable
  • Chemical compound: water based non-ionic surfactant
The concentrated formula dilutes at 1:64 for great value. It meets the Green Seal™ GS-37 Standard, including reduced health, safety, and environmental impact of formulations as well as product efficacy requirements. It also complies with LEED green building requirements and environmentally preferable product purchasing programs in place throughout federal, state, and municipal governments.