ProLysol® Advanced Deep Clean All-Purpose Cleaner, Lemon Breeze, Essendant
Supplier: Janitorial Supplies
RAC00351
76500-410CT 123.32 USD
76500-410
ProLysol® Advanced Deep Clean All-Purpose Cleaner, Lemon Breeze, Essendant
Cleaning Agents Disinfectants
Helps to deliver fresh, deep-cleaned, and disinfected surfaces.
  • Kills 99.9% of bacteria in 10 seconds when used as directed
  • Destroys tough grease and grime
  • Quickly cuts through stubborn stains and spills
The Advanced formulation powers through the toughest cleaning jobs, cutting through tough grease and soap scum. When used as directed, eliminates 99.9% of viruses and bacteria to provide peace of mind for families and facilities. It is effective against SARS-CoV-2, the virus that causes COVID-19, in 2 minutes on hard, non-porous surfaces. Suitable for use on ceramics, enamel, fiberglass, laminates, metal, plastic, and vinyl. Chemical compound: Quanternary Ammonium Compounds; Benzyl-C12-16; Alkyldimethyl; Chlrorides.