Fantastik Disinfectant Multi-Purpose Cleaner, Lemon Scent, Essendant
Supplier: Janitorial Supplies
SJN306388
76500-392CT 40.39 USD
76500-392
Fantastik Disinfectant Multi-Purpose Cleaner, Lemon Scent, Essendant
Cleaning Agents Disinfectants
Fantastik disinfectant multi-purpose cleaner, when used as directed on hard, non-porous surfaces, kills 99.9% of viruses and bacteria.
  • Fresh lemon scent neutralizes unpleasant odors
  • Fights tough grease and grime
  • Read label before use; use only as directed
Effective against germs that cause the common cold, flu, and other airborne viruses. Suitable for use on ceramics, enamel, fiberglass, glass, grout, laminates, metal, plaster, plastic, and vinyl. Chemical compound: Ammonium Chloride.