Boardwalk® Disinfecting Wipes, Lemon Scent, Essendant
Supplier: Janitorial Supplies
BWK455W75
76500-368CT 57.37 USD
76500-368
Boardwalk® Disinfecting Wipes, Lemon Scent, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Convenient cleaning wipes disinfect hard surfaces in one easy step. Bleach- and alcohol-free formula kills 99.9% of E. coli, Salmonella, Staph, and Strep. Also effective against Influenza A and H1N1 flu virus.
  • Kills 99.9% of bacteria in 15 seconds
  • Effective against SARS-CoV-2, the virus that causes COVID-19 when used as directed
  • Bleach- and alcohol-free
  • Cleans, disinfects, and deodorizes in one step
  • For use on hard, non-porous surfaces
  • Easy dispensing canister
Ready-to-use, premoistened wipes are great for offices, clinics, day care centers, restaurants, schools, and nursing homes. Wipes are packaged in an easy dispensing canister.