ChaseProd Champion Sprayon Disinfectant, Essendant
Supplier: Janitorial Supplies
CHP5157
76500-354CT 80.19 USD
76500-354
ChaseProd Champion Sprayon Disinfectant, Essendant
Cleaning Agents Disinfectants
Hospital-type formula spray disinfectant kills 99.9% of germs in 20 seconds, and 100% of germs in 10 minutes, including HIV-1 on precleaned environmental surfaces or objects.
  • Effective against fungi
  • Prevents mold and mildew on hard surfaces
Also effective against fungi and prevents mold and mildew on hard surfaces. Suitable for use on ceramics, enamel, fiberglass, laminates, plastic, metal, and vinyl. Unscented.