Rubbermaid® Pulse Microfiber Spray Mop System, Essendant
Supplier: Janitorial Supplies
RCP1835528
76500-110EA 243.57 USD
76500-110
Rubbermaid® Pulse Microfiber Spray Mop System, Essendant
Mops
Clean more square feet in less time with this reliable mop system.
  • Red ergonomic trigger handle dispenses three streams of cleaning solution with each press
  • Lightweight construction; only 4 lbs. when fully loaded with solution
  • High-capacity refillable reservoir holds 21 oz. of solution to clean up to 850 sq. ft. of floor space
  • Reservoir windows allow easy monitoring of fluid level
  • 17" wide mop head
  • 52" handle length, 56" overall length
Industry-best microfiber, on-board reservoir, and user-controlled release of solution results in faster, easier, and more effective cleaning.
System includes one trigger handle with fluid reservoir, a 17" Quick-Connect frame, and two 18" Hygen™ Damp mop pads. Also compatible with Rubbermaid® 11" and 24" Quick-Connect frames and pads.