Fecal Parasite Concentrators (FPC), Caplugs
Supplier: Caplugs
308-1000-000 308-1050-FPCSV 308-2000-000 308-4000-000
76499-416CT 154.59 USD
76499-416 76499-418 76499-420 76499-422
Fecal Parasite Concentrators (FPC), Caplugs
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
FPC Fecal Parasite Concentrator is one of the cleanest means of isolating helminth eggs and larvae, protozoan cysts, coccidian oocysts, and microsporidian spores.
  • Patented, leak-proof, self-contained two tube system
  • Standardization of concentration method, eliminating varying results among different users
  • Minimizes personnel exposure to potentially infectious specimens
  • Variety of sizes
The easy-to-use system provides clean sediment for microscopic examination and requires minimal processing time. Excellent results are obtained using either fresh or preserved specimens.
Fresh fecal specimens or specimens preserved in 5% or 10% formalin, merthiolate iodine formalin (MIF), polyvinyl alcohol (PVA) or sodium acetate-acetic acid-formalin (SAF) can be used in this system.