CE Compact Trolley, Vileda Professional®
Supplier: Vileda Professional - FHP
169843
76499-310CS 2277.11 USD
76499-310
CE Compact Trolley, Vileda Professional®
Carts
The CE Compact Trolley has been designed for use in controlled environments where footprints and becoming smaller or where storage is a concern.
  • Electropolished stainless steel for a contamination free surface
  • Autoclavable
  • Low profile design fits under work stations or tables
  • Fits full sized pre-prep boxes and buckets
  • Reduces the risk of cross contamination by staying inside the controlled space for storage
  • Accessories can be color coded
Compact, modular, and stowable trolley designed for use with pre-prepared mops and cleaning supplies, that stays inside the space and is adjustable to a variety of uses.