Cadaver Cover for JC100 Body Tray and Frame, Mortech®

Supplier: Mortech Manufacturing
JZ013
76498-638EA 373.93 USD
76498-638
Cadaver Cover for JC100 Body Tray and Frame, Mortech®
Covers
This cadaver cover is designed specifically for use with the JC100 body tray and frame.

  • For use with JC100 body tray and frame
  • Color: white

Cadaver covers allow for the discrete transport of a patient from the hospital room to the mortuary. After placing the body and the frame on the carrier, place cover over the frame. The rectangular shape of the cover conceals the body, giving the illusion of a standard gurney while hiding the cadaver.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR