BxWraps Lens Paper Wraps, Mortech®
Supplier: Mopec
BI057 BI058 BI059 BI060
76498-610PK 49.77 USD
76498-610 76498-612 76498-614 76498-616
BxWraps Lens Paper Wraps, Mortech®
Wipes Disposable Wipes and Towels Lens Papers
These blue lens paper wraps are ideal for containing and maintaining identity of small biopsy tissue. Water, alcohol and xylene resistant.
  • Water-, alcohol-, and xylene-resistant
  • 500 papers per box