20-Place Plastic Slide Holder, Mortech®
Supplier: Mortech Manufacturing
BH008
76498-594EA 80.45 USD
76498-594
20-Place Plastic Slide Holder, Mortech®
Slide Holders
Made of durable white plastic, folder holds 20 standard 3×1" slides.
  • Tranasparent cover
  • 20 slide capacity
Each slide rests on raised plastic points to prevent sticking.