Spray Lube, Mortech®
Supplier: Mortech Manufacturing
BE060
76498-478EA 45.88 USD
76498-478
Spray Lube, Mortech®
Lubricants
Spray on all moving parts of instruments after cleaning and before autoclaving to restore mobility and range of instrument motion.
  • 8 fluid oz. container
Instruments must be autoclaved prior to spraying. Allow instrument to drain before applying lube. Rinsing and wiping are unnecessary after application.