SaniPath Disinfecting Spray Cleaner, Mortech®
Supplier: Mortech Manufacturing
BE047
76498-466EA 40.54 USD
76498-466
SaniPath Disinfecting Spray Cleaner, Mortech®
Cleaning Agents Disinfectants
Protect personnel and their work areas with SaniPath Disinfecting Spray, a multi-purpose decontaminating cleaner. It is a bactericidal, fungicidal, virucidal, and Tuberculocide.
  • Mild, lemon fragrance leaves the lab smelling fresh
  • 32 oz. container
This cleaner is great for spill clean up and should be used daily for pathology and morgue work surfaces including equipment, tables, coolers, cutting stations, instrumentation, floors, computers, and telephones.