Eazy Soak, Instrument Cleaner, Mortech®
Supplier: Mortech Manufacturing
BE046
76498-464EA 121.53 USD
76498-464
Eazy Soak, Instrument Cleaner, Mortech®
Cleaning Agents Disinfectants
This unique, clear, odorless enzyme instrument cleaner is ideal for an overnight easy soak.
  • No scrubbing or pre-rinsing is required
  • 32 oz. container
Ideal for hard-to-clean instruments such as scissors, forceps, serrated cutting blades, and scalpel handles. It digests all tissue, blood, and exudative material.