SaniPath Disinfectant Foaming Spray, Mortech®
Supplier: Mortech Manufacturing
BE045
76498-462EA 40.84 USD
76498-462
SaniPath Disinfectant Foaming Spray, Mortech®
Cleaning Agents Disinfectants
SaniPath foaming spray destroys a wide range of pathogenic bacteria, viruses, and fungi, including HIV-1, HIN1, Herpes Simplex 2, Staphylococcus, Pseudomonas, Canine Parvo, and Tuberculosis.
  • Use at least once a week to keep labs safe and clean
  • 32 oz. container
Clean, disinfect and deodorize in 10 minutes with the clinging foam. The powerful foaming action is ideal for all heavily soiled vertical or horizontal pathology surfaces such as autopsy tables, body trays, grossing stations, plumbing fixtures, and dissection boards.