PathOdorizer, Odor Eliminating Solution, Mortech®
Supplier: Mortech Manufacturing
BE040 BE042
76498-458EA 120.64 USD
76498-458 76498-460
PathOdorizer, Odor Eliminating Solution, Mortech®
Cleaning Agents Detergents and Surface Cleansers
Developed specially for use by medical examiners, PathOdorizer eliminates offensive odors emitted from decomposed bodies within seconds. From the discovery scene to the morgue, PathOdorizer can be used in every phase of work.
  • No CFCs, formaldehyde, or hazardous biocides
  • No toxic fumes
  • Safe for use on metals, plastics, or fabrics
  • PathOdorizer Plus offers additional cleaning and degreasing properties
PathOdorizer not only removes odors from clothes, vehicles, body bags, refrigerators, and other items, it keeps the odor from permeating the lab environment. The formula is completely water soluble, odorless, colorless, biodegradable, non-flammable, and non-corrosive.