Hydro Aspirator Extension, Mortech®
Supplier: Mortech Manufacturing
BE015
76498-434EA 329.84 USD
76498-434
Hydro Aspirator Extension, Mortech®
Bottles Aspirators
This hydro-aspirator extension is fabricated from chrome plated brass. It provides further safety against polluted backflow.
Ordering information: Unit can be purchased individually, or part of a whole water control unit.