Autopsy Head Drapes and Framework, Mortech®
Supplier: Mortech Manufacturing
BE010 BE011
76498-426EA 571.45 USD
76498-426 76498-428
Autopsy Head Drapes and Framework, Mortech®
First Aid Supplies Surgical Drapes
Drapes help to contain infectious and contaminated materials during autopsy procedures.
  • Easily assembled
  • Use for a variety of head, torso, and/or appendage dissections
Head drapes are made from 0.03 mm thick polyethylene. The drapes are not autoclavable.