Autopsy Saw Blades for Mopec Swordfish Autopsy Saw, Mortech®
Supplier: Mortech Manufacturing
BD140 BD141 BD142 BD143
76498-398EA 231.37 USD
76498-398 76498-400 76498-402 76498-404
Autopsy Saw Blades for Mopec Swordfish Autopsy Saw, Mortech®
Razors and Blades
Round, sectional, and segmented blades are available for any bone cutting application, including cranial removals, cross and long bone sectioning, and vertebral column.
  • Choose from section blade, round blade, large round blade, or spinal column blade
  • Stainless steel construction
Axial mount blades eliminate nicks to fingers and knuckles.