Amputation Dissecting Board, Mortech®
Supplier: Mortech Manufacturing
BC017
76498-304EA 1097.04 USD
76498-304
Amputation Dissecting Board, Mortech®
Dissecting Boards
Designed by Pathologist David Pinkhasov, this uniquely designed product offers safe and secure dissection of any amputated limb specimen.
  • 15" (38 cm) height
  • Polyethylene construction with stainless steel clamps
  • Opaque white finish
The Amputation dissection board securely sits on any table surface and features heavy gauge stainless steel clamps to hold any amputation specimen in place.