Stainless Steel Dissecting Tray, Mortech®
Supplier: Mortech Manufacturing
BC010
76498-300EA 558.04 USD
76498-300
Stainless Steel Dissecting Tray, Mortech®
Trays/Dishes/Pots Dissecting Trays
A multi-purpose unit, this durable tray can be used to confine spills or contamination to a restricted area.
  • All stainless steel
  • Dissecting tray with sloped angle end for draining
All of Mopec’s standard dissecting boards will fit into this 1" deep tray. The sloped end overhangs any sink, allowing all fluids to drain directly into the sink.