Standard and Premium Biopsy Bags, Mortech®
Supplier: Mopec
BBS-1000-01 BBM200-02 BBL500-02 BBM200-01 BBS1000-02
29000-048PK 4678.43 USD
29000-048 29000-050 76498-276 76498-278 76498-282
Standard and Premium Biopsy Bags, Mortech®
Bags Sample Bags
Biopsy bags are available in three standard sizes and two premium sizes.
  • Biopsy bags are made of quality nylon
  • Reduced risk of small specimen loss
Bags peel open for quick and easy specimen removal. The design allows unrestricted fluid movement around tissue.