Washdown Bench Scale, Mortech
Supplier: Mortech Manufacturing
BB027
76498-258EA 5241.98 USD
76498-258
Washdown Bench Scale, Mortech
Balances and Scales
Constructed of a stainless steel base with an ABS indicator. Results quickly display on large backlit LCD screen. Four key membrane keypad with raised tactile keys.
  • LCD 11 mm display
  • kg, g, lb. and user definable custom units
  • 10,000:12,500 default resolution, 1:5000 NTEP Certified Resolution
  • Includes RS232 interface with GMP/GLP data output including date and time
Weight: 29.04 lbs.
Accessories information: A rechargeable battery kit is available for use with BB027 scale.