Stainless Steel Headrest, Mortech®
Supplier: Mortech Manufacturing
BA078
76498-240EA 151.46 USD
76498-240
Stainless Steel Headrest, Mortech®
Ergonomic Supports Shoulder Supports
Designed to provide a more natural cranial positioning during autopsy and mortuary procedures.
  • 13.75" length
  • Stainless steel construction
  • Four non-slip foot pads for stability
The broad convex curve fits within the arch of the neck, allowing the cadaver head to tilt up and rest upon a work surface. This configuration allows easy access to the neck and ears without interference.