Wedge-Ease Body Support, Mortech®
Supplier: Mortech Manufacturing
BA029
76498-200EA 322.07 USD
76498-200
Wedge-Ease Body Support, Mortech®
Ergonomic Supports Back Supports
This uniquely shaped post-mortuary support tool is designed to help isolate, shift, and/or secure a body during autopsy or embalming procedures.
  • For use under legs, buttocks, back, and arms
  • Support will grip any table
  • Chemically-resistant