VWR® pHenomenal® MU 6100 H Handheld Meters (pH/conductivity/DO)

Supplier: VWR International
2FV320MOY 2FV321 2FV322
76460-522EA 1807.56 USD
76460-522 76498-140 76498-142
VWR® pHenomenal® MU 6100 H Handheld Meters (pH/conductivity/DO)
Electrochemistry Meters pH and Multi-Parameter Meters
This user-friendly, portable instrument offers high resolution and accurate precision measurements for pH, mV, conductivity, and oxygen in on-site applications. It can measure a maximum of 2 of these parameters simultaneously.

  • Large graphic LCD with continuous LED backlight showing both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets; additional 200 memory points with manual storage
  • Automatic power shut-off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1.68/4.00/6.86/9.18/12.54)
  • Three additional technical buffers at 25 °C (4.00/7.00/10.00) in memory
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
  • Low energy consumption: Choice of mains supply or 4× AA batteries for over 1000 operating hours

With features that include waterproof USB output for connection to a computer and a rugged IP 67 enclosure.

Certifications: GLP compliant.

Ordering information: Conductivity sensors for low conductivities are available on request. For more information, please contact your Avantor representative.

Delivery information: The ready-to-use kit MU 6100 H S1 (76498-140) comes with a multi- parameter measuring device in a robust plastic case, pH gel electrode 111 made of epoxy with integrated temperature sensor (1 m cable), starter kit (two pH buffers, one plastic cup), Conductivity sensor CO 11 with integrated temperature sensor (1.5 m cable), stand and operating instructions (22 languages) supplied on CD-ROM.
The MU 6100 H S2 kit (76498-142) also includes the OXY 11-3 galvanic oxygen sensor with an integrated temperature sensor (3 m cable) and a maintenance kit.
76460-522 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR