Linearity FLQ Urine/Fluids Chemistry for Beckman AU, AUDIT MircoControls
Supplier: AUDIT MICROCONTROLS
K828M-5
76492-630EA 311.08 USD
76492-630
Linearity FLQ Urine/Fluids Chemistry for Beckman AU, AUDIT MircoControls
Controls and Calibration Verification Materials
The Audit® MicroControlsTM Linearity FLQ Urine/Fluids Chemistry for Beckman AU is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, Sodium, Phosphorous, Urea Nitrogen, Uric Acid.