InMotion KF Sample Vials
Supplier: Mettler Toledo
30407445
76492-556PK 239 USD
76492-556
InMotion KF Sample Vials
Vials
These clear glass sample vials are used with InMotion KF oven autosampler.
  • Made of hydrolytic class 1 borosilicate glass
  • Withstand temperatures from −20 to +121 °C
These vials are suitable for sample storage and analysis in a variety of laboratory applications.