Universal Transport Medium (UTM®) Mini-tip Flocked Swab with Breakpoint, No Beads
Supplier: Hardy Diagnostics
3C057N
76492-464EA 1611.08 USD
76492-464
Universal Transport Medium (UTM®) Mini-tip Flocked Swab with Breakpoint, No Beads
Swabs Transport Swabs
UTM® is a FDA cleared collection and transport system suitable for collection, transport, maintenance, and long-term freeze storage of clinical specimens containing viruses, including COVID-19, chlamydia, mycoplasma, or ureaplasma organisms. The transport medium comes in a plastic, screw cap tube and maintains organism viability for 48 hours at room or refrigerated temperature.
  • Room temperature stable system maintains viability of organisms for up to 48 hours
  • Does not require refrigeration storage prior to collection
  • Skirted tube
  • Rapid release and
More Product Information