VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Supplier: VWR International
76491-688
76491-688CS 323.67 USD
76491-688
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Timers Digital Timers
Designed with 10 buttons, this timer allows direct input for faster programming of desired time.
  • Counts up and down, and displays elapsed time after countdown
  • Capacity of 99 minutes, 99 seconds
  • Audible and visual alarms
  • Last time set memory
  • Requires two AAA batteries
Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment.
Certifications: Individually tested and inspected using NIST in A2LA accredited laboratory (certification #2448.01), ensuring conformity to ISO 17025:2005. RoHS compliant.
Delivery information: Supplied with flip stand, magnetic back, pocket clip, two AAA batteries, and certificate of calibration.