VWR® Single Channel Switchable Electronic Timer
Supplier: VWR International
76491-686
76491-686CS 931.74 USD
76491-686
VWR® Single Channel Switchable Electronic Timer
Timers Digital Timers
This single-channel timer is switchable between 19 hours, 59 minutes and 99 minutes, 59 seconds. It features a 3-key operation for ease of use.
  • Count direction: up/down
  • Last time set memory
  • Audible alarm; displays elapsed time after countdown
Mounting options: lay flat on bench, flip-open stand for vertical view, and a magnetic back for metal surface attachment.
Certifications: Certificate of calibration and individual serial number, these electronic timers are tested and calibrated in an exclusive triple-accredited/-registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Supplied with fold-away stand, pocket clip, magnetic back, one AAA battery, and certificate of calibration. Includes a statement of traceability indicating accuracy to NIST.