VWR® Freezer Verification Thermometer
Supplier: VWR International
76491-664
76491-664CS 665.72 USD
76491-664
VWR® Freezer Verification Thermometer
Thermometers Analog Thermometers
This mercury free thermometer is traceable to NIST and covers a range from −25 to −5 °C. These thermometers are suitable for applications with freezers. The thermometer is placed in a bottle filled with a liquid.
  • Verify sample temperatures in freezers
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Thermometer contains Enviro-Safe® liquid
  • Has permanently fused markings and lead free glass
  • PFA coating safely contains glass and liquid in case of breakage
  • Individually serialized
Insert the thermometer into the bottle and place near the samples for best results.
Certifications: Traceable to NIST.