VWR® General Purpose Liquid-In-Glass Thermometer, Red Spirit
Supplier: VWR International
76491-636
76491-636CS 589.55 USD
76491-636
VWR® General Purpose Liquid-In-Glass Thermometer, Red Spirit
Thermometers Analog Thermometers
These thermometers contain red organic liquid against white back glass and are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries.
  • Total immersion
  • Red liquid against white back glass
  • Permanent scale embedded in the glass