VWR® Environmentally Friendly Thermometers, Easy Read
Supplier: VWR International
76491-596 76491-598 76491-600 76491-602 76491-604 76491-606
76491-596CS 542.69 USD
76491-596 76491-598 76491-600 76491-602 76491-604 76491-606
VWR® Environmentally Friendly Thermometers, Easy Read
Thermometers Analog Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Eliminates reading errors
  • Non-toxic, non-hazardous, biodegradable liquid
Includes multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability, non roll fitting and individual serial number.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).