Air Ionizing Gun, Transforming Technologies
Supplier: Transforming Technologies
IN6200
76489-744EA 1432.8 USD
76489-744
Air Ionizing Gun, Transforming Technologies
Ionizers
A lightweight but durable static removal gun.
  • Point and shoot precision ionization
  • Durable impact-resistant fiberglass reinforced case
  • Quick disconnect air fittings
The ionizing gun provides superior control and targeted particulate cleanup for a wide variety of industrial applications. As a forceful neutralizer and blow-off gun, it easily removes stubborn particulate from surfaces. As a neutralizer, it gets rid of troublesome charges responsible for the attraction of contaminants to product surfaces.