Disposable Heel Grounder, Transforming Technologies
Supplier: Transforming Technologies
HG4560L
76489-732PK 99.12 USD
76489-732
Disposable Heel Grounder, Transforming Technologies
Grounders Heel and Toe Grounders
Disposable heel grounders are comfortable to wear, easily identifiable, and use the entire volume of the fabric to provide protection.
  • Low cost method of grounding temporary personnel
  • High visibility yellow color
  • Disposable design is perfect for one time use
Constructed of a durable non-woven fabric impregnated with carbon, these disposable heel grounders use the entire volume of the fabric to provide protection.