Heel Grounder, with 1.25" Cup and Buckle Closure, Transforming Technologies
Supplier: Transforming Technologies
HG1380
76489-724EA 7.2 USD
76489-724
Heel Grounder, with 1.25" Cup and Buckle Closure, Transforming Technologies
Grounders Heel and Toe Grounders
Constructed from durable materials with a 0.75" buckle closure to maximize fit and comfort.
  • Buckle closure
  • Buried 1 meg resistor for safety
  • Meets or exceeds requirements of ANSI ESD-S20.20
Heel grounding devices provide protection for staff on the move.