Universal Snap Kit, Transforming Technologies
Supplier: Transforming Technologies
CS1090
76489-640PK 21.42 USD
76489-640
Universal Snap Kit, Transforming Technologies
Grounders Grounding Wires
Universal snap kit easily installs a ground snap into a range of mat thicknesses.
  • Install with a screwdriver
  • Installs a 10 mm snap or stud to ESD workstation materials
  • Includes two screws for thick and thin materials
Fast and easy way to install 10 mm snaps or studs to soft work station materials. Includes two screws (floor and table mat lengths), and one each of 10 mm socket, 10 mm stud, washer, and screw base.