Molded Banana Jack Snap, 10 mm Stud and Socket, Transforming Technologies
Supplier: Transforming Technologies
CS0127
76489-634PK 42.84 USD
76489-634
Molded Banana Jack Snap, 10 mm Stud and Socket, Transforming Technologies
Grounders Grounding Wires
The Molded Banana Jack Snap adds a banana jack to a table mat.
  • Add a banana jack to any surface with a 10 mm ground snap
  • Molded plastic body with metal snaps
  • Use with coil cords, wrist strap sets, common point grounds, and constant monitors
Fully molded in plastic, the device has both male and female 10 mm snaps with a banana jack, and is compatible and interchangeable with standard banana plugs. It can connect to a wrist strap, ground cord, or constant monitor, and is excellent for grounding carts.