IDP-STAT® ESD Badge Holder, Transforming Technologies
Supplier: Transforming Technologies
5221.3.IDP
76489-526PK 28.1 USD
76489-526
IDP-STAT® ESD Badge Holder, Transforming Technologies
Personal Identification ID Card Holders
ESD Badge Holders are an excellent way to shield employee badges, guest passes, or key cards. The ESD protection makes them safe for use in sensitive environments.
  • Permanently dissipative
  • Overall dimensions: 3.75×3.25"
  • Fits most IDs
The IDP-STAT® ESD Badge Holders are made from permanently dissipative IDP-STAT® PVC film. These badge holders will never fail in the field and become a potential ESD threat.