LiquidGuard™ Sterile Sure-Clean Sleeve Covers, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
SGR1919LGWH21T-200
76488-320CS 319.22 USD
76488-320
LiquidGuard™ Sterile Sure-Clean Sleeve Covers, Apex Aseptic Products
Disposable Garments
Sterile LiquidGuard™ sleeve covers are suitable for cleanroom use.
  • LiquidGuard™ microporous fabric construction
  • Impervious yet breathable
  • Easy breathability, comfortable to wear
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Latex-free, TSE-free, BSE-free
  • Manufactured in a controlled environment
  • Gamma irradiated validated sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Sleeves feature attached thumb loops to help keep them in place. Tunneled sewing greatly reduces particle shedding. A special folding system helps to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670.
Delivery information: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Sleeves are packaged 2 pieces per package, triple bagged, 200 pieces per case.