VWR® Plus Series Undercounter Laboratory Freezers Freestanding (–20 °C)
Supplier: VWR International
76488-176
76488-176EA 2071.59 USD
76488-176
VWR® Plus Series Undercounter Laboratory Freezers Freestanding (–20 °C)
Freezers Laboratory / General Purpose Freezers
The compact undercounter laboratory freezer provides quality cold storage in limited-space settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (⅜")
  • HFC-free refrigerant (R600a)
This compact, freestanding undercounter freezer offers a cold-storage solution for limited space in laboratory settings. The unit offers premium features for laboratory storage including a series of temperature alarms to keep stored products at peak effectiveness. Units are equipped with keyed door locks, LED interior lighting, a probe access port, and a microprocessor temperature controller.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. These freezers run on natural, hydrocarbon refrigerant for environmental cleanliness and economic efficiency.
Material: White cabinet and door
Certifications: UL/C-UL listed