SP Bel-Art Touch Free™ Automatic Waste Cans, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13202-0050
74950-066EA 352.35 USD
74950-066 89125-226 76486-082
SP Bel-Art Touch Free™ Automatic Waste Cans, Bel-Art Products, a part of SP
Bins Waste Bins
The Touch Free™ Automatic Waste Can’s motion-sensitive lid opens automatically to accept waste, and then closes into a fitted well that helps to reduce odor and airborne contamination.
  • Durable and dependable
  • On-off switch deactivates the auto-open/close function
  • Up to 4000 openings per set of batteries (not included)
Use with SP Bel-Art super-strength or standard red Biohazard disposal bags, clear Biohazard disposal bags, Clavies® transparent autoclavable bags, or commercially available waste bags.
Delivery information: Supplied with a pre-printed, brightly colored Biohazard label, 2 others for Recyclables and Trash, and a blank label for individualized applications.