Immobilized T4 DNA Ligase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0569S
76486-074EA 318 USD
76486-074
Immobilized T4 DNA Ligase, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Immobilized T4 DNA Ligase (IM-T4 DNA Ligase) is a slurry of magnetic beads coated with T4 DNA Ligase to produce a 10 mg per ml solution (50% glycerol) with an effective concentration of 60 cohesive end units (CEU) per microliter of slurry. Following a reaction, enzyme can be removed using a magnet thereby avoiding heat inactivation, and can be re-used.
  • Easily remove enzyme from reaction using a magnet
  • Avoid heat inactivation step, which may interfere with downstream applications
  • Reusable up to 20 times
  • Enable new workflows and applications