K5 Monochrome sCMOS Camera, Leica Microsystems
Supplier: Leica Microsystems
11547112
76485-962EA 8912.23 USD
76485-962
K5 Monochrome sCMOS Camera, Leica Microsystems
Cameras
For routine fluorescence imaging.
  • Bring the flexibility and power of sCMOS technology into your lab and image the details of your experiment
  • Efficiently segment crucial, real-time events
  • Improve signal-to-background of your THUNDER imager
Image a wide variety of sample types with the flexibility and power of high 4.2 megapixel resolution and fast 40 fps imaging brought to you by the K5. Recommended for live imaging and lower signal-to-noise fluorescence imaging, especially when paired with a Leica THUNDER imager.