Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K950M-5
76485-888EA 381.4 USD
76485-888
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Standards Chromatography Standards
The Audit® MicroControls™ Linearity FLQ TDM for Abbott systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Acetominophen, Amikacin, Carbamezepine, Digoxin, Gentamicin, Phenobarbital, Phenytoin, Salicylate, Theophylline, Tobramycin, Valproic Acid, Vancomycin.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.