iFluor® 665, Maleimide fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1554
76485-762EA 230 USD
76485-762
iFluor® 665, Maleimide fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbb14cc16b6c8932eb3f2a052b42c1caed16376d&vid=a8799b214a9ff00b7a456a46454fd76068c93e7c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR